Pierce™ Protein A/G Plus Agarose, 20 mL - FAQs
Pierce™ Protein A/G Plus Agarose, 20 mL - FAQs
View additional product information for Pierce™ Protein A/G Plus Agarose - FAQs (20423, 20424)
2 product FAQs found
I have noticed that there is a slightly higher amount of agarose resin in product Cat. No. 20424 than expected according to the manual. What is the reason for that?
To ensure the delivery of the correct amount of agarose resin, the products are slightly overfilled with resin. The slurry will be maintained at 50% so the resin and liquid solution volume will be equivalent.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What does it mean that the crosslinked 6% beaded agarose is supplied as a 50% slurry for products Pierce Protein A/G Plus Agarose (Cat. No. 20423 and 20424) and Pierce Protein A/G Agarose (Cat. No. 20421 and 20422)?
This means that the solution consists of 50% settled resin and 50% liquid solution. For example, Cat. No. 20424 contains 20 ml of settled resin and 20 ml of storage buffer, making it a 40 ml, 50% slurry.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.