5-Bromopyrimidine, 98%
5-Bromopyrimidine, 98%
5-Bromopyrimidine, 98%
Thermo Scientific Chemicals

5-Bromopyrimidine, 98%

CAS: 4595-59-9 | C4H3BrN2 | 158.986 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 204301000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4595-59-9
IUPAC Name5-bromopyrimidine
Molecular FormulaC4H3BrN2
InChI KeyGYCPLYCTMDTEPU-UHFFFAOYSA-N
SMILESBrC1=CN=CN=C1
さらに表示
Melting point69°C to 73°C
Appearance (Color)Yellow to beige to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder and/or chunks
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only