N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%
N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%
N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%
Thermo Scientific Chemicals

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%

CAS: 3575-32-4 | C8H12N2·2HCl | 209.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 204340050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3575-32-4
IUPAC NameN1,N1-dimethylbenzene-1,3-diamine dihydrochloride
Molecular FormulaC8H14Cl2N2
InChI KeyBZJPIQKDEGXVFG-UHFFFAOYSA-N
SMILESCl.Cl.CN(C)C1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey to purple
Titration Argentometric>=98.5 %
Appearance (Form)Powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only