Eicosane, 99%
Eicosane, 99%
Eicosane, 99%
Thermo Scientific Chemicals

Eicosane, 99%

CAS: 112-95-8 | C20H42 | 282.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 204371000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-95-8
IUPAC Nameicosane
Molecular FormulaC20H42
InChI KeyCBFCDTFDPHXCNY-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
Melting point36°C to 38°C
Infrared spectrumConforms
Appearance (Color)Transparent white
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only