DL-2,3-Diaminopropionic acid monohydrochloride, 99%
DL-2,3-Diaminopropionic acid monohydrochloride, 99%
DL-2,3-Diaminopropionic acid monohydrochloride, 99%
Thermo Scientific Chemicals

DL-2,3-Diaminopropionic acid monohydrochloride, 99%

CAS: 54897-59-5 | C3H8N2O2·HCl | 140.57 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 204670250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS54897-59-5
IUPAC Namehydrogen 2,3-diaminopropanoic acid chloride
Molecular FormulaC3H9ClN2O2
InChI KeySKWCZPYWFRTSDD-UHFFFAOYNA-N
SMILES[H+].[Cl-].NCC(N)C(O)=O
さらに表示
Appearance (Color)White to light beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only