DL-Homoserine, 99%
DL-Homoserine, 99%
DL-Homoserine, 99%
Thermo Scientific Chemicals

DL-Homoserine, 99%

CAS: 1927-25-9 | C4H9NO3 | 119.12 g/mol
Have Questions?
Catalog NumberQuantity
2047600505 g
Catalog number 204760050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1927-25-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
Titration with HClO498.5 to 101.0 %
Infrared spectrumConforms
Appearance (Color)White
Solubility(1 % in water) Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only