Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Thermo Scientific Chemicals

Sodium phosphate, dibasic, 99+%, for analysis, anhydrous

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 204850050
Price (USD)
-
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or granular powder
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
Sulfate (SO4)=<50 ppm
Loss on drying=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
RUO – Research Use Only