Iminodiacetic acid, 98+%
Iminodiacetic acid, 98+%
Iminodiacetic acid, 98+%
Thermo Scientific Chemicals

Iminodiacetic acid, 98+%

CAS: 142-73-4 | C4H7NO4 | 133.1 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 g
2.5 kg
產品號碼 204975000
價格 (TWD)
-
Quantity:
500 g
Request bulk or custom format
化學識別
CAS142-73-4
IUPAC Name2-[(carboxymethyl)amino]acetic acid
Molecular FormulaC4H7NO4
InChI KeyNBZBKCUXIYYUSX-UHFFFAOYSA-N
SMILESOC(=O)CNCC(O)=O
檢視更多
規格Specification Sheet規格表
Titration with NaOH>=98.0 %
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only