Iron(II) chloride tetrahydrate, 99+%
Iron(II) chloride tetrahydrate, 99+%
Iron(II) chloride tetrahydrate, 99+%
Iron(II) chloride tetrahydrate, 99+%
Thermo Scientific Chemicals

Iron(II) chloride tetrahydrate, 99+%

CAS: 13478-10-9 | Cl2Fe·4H2O | 198.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 205082500
Price (EUR)
50,65
Online Exclusive
56,30
Save 5,65 (10%)
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
50,65
Online Exclusive
56,30
Save 5,65 (10%)
Each
Chemical Identifiers
CAS13478-10-9
IUPAC Nametetrahydroxyironbis(ylium) dichloride
Molecular FormulaCl2FeH8O4
InChI KeyWSSMOXHYUFMBLS-UHFFFAOYSA-L
SMILESO.O.O.O.[Cl-].[Cl-].[Fe++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green
Appearance (Form)Crystalline powder, crystals and/or chunks
Titration with KMnO4>=99.0 %
Sulfate (SO4)=<200 ppm
Arsenic (As)=<5 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only