D(-)-Lyxose, 99+%, mixture of anomers
D(-)-Lyxose, 99+%, mixture of anomers
D(-)-Lyxose, 99+%, mixture of anomers
Thermo Scientific Chemicals

D(-)-Lyxose, 99+%, mixture of anomers

CAS: 1114-34-7 | C5H10O5 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 205230050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1114-34-7
IUPAC Name2,3,4,5-tetrahydroxypentanal
Molecular FormulaC5H10O5
InChI KeyPYMYPHUHKUWMLA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Specific optical rotation-13° to -15° (20°C, 589 nm) (c=2, H2O)
Infrared spectrumConforms
Appearance (Color)White to light yellow
HPLC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only