Mercury(II) iodide, red, 99+%
Mercury(II) iodide, red, 99+%
Mercury(II) iodide, red, 99+%
Thermo Scientific Chemicals

Mercury(II) iodide, red, 99+%

CAS: 7774-29-0 | HgI2 | 454.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 205740025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7774-29-0
IUPAC Namemercury(2+) diiodide
Molecular FormulaHgI2
InChI KeyYFDLHELOZYVNJE-UHFFFAOYSA-L
SMILES[I-].[I-].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Soluble Hg salts (as Hg)=<0.05 %
Appearance (Color)Red
Titration Iodometric>=99.0 %
Mercurous Mercury (as Hg)=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only