Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis
Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis
Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis

CAS: 12054-85-2 | H24Mo7N6O24·4H2O | 1235.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 205850025
Price (USD)
529.65
Online exclusive
588.00
Save 58.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
529.65
Online exclusive
588.00
Save 58.35
Each
Chemical Identifiers
CAS12054-85-2
IUPAC Nameammonium hydrate molybdenum oxidandiide
Molecular FormulaH6MoNO2
InChI KeyXCOMAVCHADVANQ-UHFFFAOYSA-O
SMILES[NH4+].O.[O--].[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Heavy metals (as Pb)=<10 ppm
Chloride (Cl)=<10 ppm
Phosphate (PO4)=<5 ppm
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
RUO – Research Use Only