Ammonium nitrate, 99+%, for analysis
Ammonium nitrate, 99+%, for analysis
Ammonium nitrate, 99+%, for analysis
Thermo Scientific Chemicals

Ammonium nitrate, 99+%, for analysis

Ammonium nitrate, 99+%, H4N2O3, CAS Number-6484-52-2, ammonium nitricum, ammonium saltpeter, merco prills, herco prills, nitric acid ammonium salt, nitram, nitrato amonico, nitrate of ammonia, german saltpeter, ammonium nitrate, 500g, 210.0 deg.C, CHEBI:63038, 5ppm max, 5ppm max., White, 1.7200g/mL | CAS: 6484-52-2 | H4N2O3 | 80.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
205861000100 gPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 205861000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS6484-52-2
IUPAC Nameammonium nitrate
Molecular FormulaH4N2O3
InChI KeyDVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES[NH4+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Adhering crystals
pH4.5 to 6 (5 % at 20°C)
Iron (Fe)=<2 ppm
Heavy metals (as Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only