Ammonium sulfate, 99.5%, for analysis, Thermo Scientific Chemicals
Ammonium sulfate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium sulfate, 99.5%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
250 g
1 kg
5 kg
Packaging:
View all
Fibre drum
Catalog number 205870250
Price (USD)/ Each
-
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Titration excess NaOH>=99.5 %
Iron (Fe)=<5 ppm
Appearance (Form)Crystals
Residue after ignition=<0.01 % (as SO4)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only