Manganese(II) chloride tetrahydrate, 99+%, for analysis
Manganese(II) chloride tetrahydrate, 99+%, for analysis
Manganese(II) chloride tetrahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Manganese(II) chloride tetrahydrate, 99+%, for analysis

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 205891000
Price (USD)
52.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
52.00
Each
Chemical Identifiers
CAS13446-34-9
IUPAC Namemanganese(2+) tetrahydrate dichloride
Molecular FormulaCl2H8MnO4
InChI KeyCNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILESO.O.O.O.[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
Sulfate (SO4)=<50 ppm
Iron (Fe)=<5 ppm
Magnesium (Mg)=<50 ppm
KMnO4-reducing substance=<0.0005 % (as O)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only