Potassium nitrate, 99+%, for analysis
Potassium nitrate, 99+%, for analysis
Potassium nitrate, 99+%, for analysis
Thermo Scientific Chemicals

Potassium nitrate, 99+%, for analysis

CAS: 7757-79-1 | KNO3 | 101.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 205910010
Price (EUR)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7757-79-1
IUPAC Namepotassium nitrate
Molecular FormulaKNO3
InChI KeyFGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES[K+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Iodate (IO3)=<5 ppm
Sodium (Na)=<200 ppm
Appearance (Form)Crystals
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only