Potassium phosphate, monobasic, 99+%, extra pure
Potassium phosphate, monobasic, 99+%, extra pure
Potassium phosphate, monobasic, 99+%, extra pure
Thermo Scientific Chemicals

Potassium phosphate, monobasic, 99+%, extra pure

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 205920100
Price (USD)
493.00
Each
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
493.00
Each
Chemical Identifiers
CAS7778-77-0
IUPAC Namepotassium dihydrogen phosphate
Molecular FormulaH2KO4P
InChI KeyGNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES[K+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=99.0 % (on dried substance)
pH4.2 to 4.7 (1 % at 20°C)
Appearance (Form)Crystalline powder or crystals
Loss on drying=<1.0 % (105°C)
Arsenic (As)=<3 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only