Sodium tetraborate decahydrate, 99.5%, for analysis
Sodium tetraborate decahydrate, 99.5%, for analysis
Sodium tetraborate decahydrate, 99.5%, for analysis
Thermo Scientific Chemicals

Sodium tetraborate decahydrate, 99.5%, for analysis

CAS: 1303-96-4 | B4Na2O7·10H2O | 381.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 205950050
Price (EUR)
245,00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
245,00
Each
Chemical Identifiers
CAS1303-96-4
IUPAC Namedisodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
Molecular FormulaB4H20Na2O17
InChI KeyOJPFLODMGUZMKD-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
pH9 to 9.5 (5 % at 20°C)
Appearance (Color)White
Heavy metals (as Pb)=<10 ppm
Chloride (Cl)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
RUO – Research Use Only