1-Octanesulfonic acid, sodium salt monohydrate, 98%
1-Octanesulfonic acid, sodium salt monohydrate, 98%
1-Octanesulfonic acid, sodium salt monohydrate, 98%
Thermo Scientific Chemicals

1-Octanesulfonic acid, sodium salt monohydrate, 98%

CAS: 207596-29-0 | C8H19NaO4S | 234.29 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 206140250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS207596-29-0
IUPAC Namesodium hydrate octane-1-sulfonate
Molecular FormulaC8H19NaO4S
InChI KeyMBURIAHQXJQKRE-UHFFFAOYSA-M
SMILESO.[Na+].CCCCCCCCS([O-])(=O)=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Powder
Titration after Ion exchange>=97.5 %
Loss on drying7 % to 9 % (140°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only