Is Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50 (Cat. No. 206231000, 206230025) supplied in weight or atomic ratio?
Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50 (Cat. No. 206231000, 206230025) is supplied in weight.
Search
View additional product information for Aluminum-nickel, Raney-type alloy, powder, Al-Ni 50/50 - FAQs (206230025, 206231000)
1 product FAQs found