Molybdenum(VI) oxide, 99+%
Molybdenum(VI) oxide, 99+%
Molybdenum(VI) oxide, 99+%
Thermo Scientific Chemicals

Molybdenum(VI) oxide, 99+%

CAS: 1313-27-5 | MoO3 | 143.95 g/mol
数量:
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 206361000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1313-27-5
IUPAC Nametrioxomolybdenum
Molecular FormulaMoO3
InChI KeyJKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILESO=[Mo](=O)=O
さらに表示
Appearance (Color)White to yellow to green to grey
Appearance (Form)Powder
PO4, AsO4, SiO4 (as PO4)=<0.002 %
Assay>=99.0 %
Sulfate (SO4)=<200 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only