Molybdenum(VI) oxide, 99+%
Molybdenum(VI) oxide, 99+%
Molybdenum(VI) oxide, 99+%
Molybdenum(VI) oxide, 99+%
Thermo Scientific Chemicals

Molybdenum(VI) oxide, 99+%

CAS: 1313-27-5 | MoO3 | 143.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 206365000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1313-27-5
IUPAC Nametrioxomolybdenum
Molecular FormulaMoO3
InChI KeyJKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILESO=[Mo](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to green to grey
Appearance (Form)Powder
PO4, AsO4, SiO4 (as PO4)=<0.002 %
Assay>=99.0 %
Sulfate (SO4)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only