Phosphomolybdic acid hydrate, 80%
Phosphomolybdic acid hydrate, 80%
Phosphomolybdic acid hydrate, 80%
Thermo Scientific Chemicals

Phosphomolybdic acid hydrate, 80%

CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 206381000
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS51429-74-4
IUPAC Name12-Molybdophosphoric acid
Molecular FormulaH3Mo12O40P
Molecular Weight (g/mol)1825.40
Synonymphosphoric acid dodecamolybdenum hexatriacontaoxidandiide
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<200 ppm
Iron (Fe)=<50 ppm
Sulfate (SO4)=<250 ppm
Appearance (Color)Yellow to yellow-orange
Assay>=47.5 % (Mo)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only