Ammonium iodide, 99+%, for analysis
Ammonium iodide, 99+%, for analysis
Ammonium iodide, 99+%, for analysis
Thermo Scientific Chemicals

Ammonium iodide, 99+%, for analysis

CAS: 12027-06-4 | H4IN | 144.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 206461000
Price (EUR)
72,70
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
72,70
Each
Chemical Identifiers
CAS12027-06-4
IUPAC Nameammonium iodide
Molecular FormulaH4IN
InChI KeyUKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES[NH4+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
AppearanceMay turn yellow on storage
Titration Argentometric99.0 to 101.0 %
Iron (Fe)=<5 ppm
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only