ART™ Barrier Pipette Tips in Hinged Racks
Thermo Scientific™

ART™ Barrier Pipette Tips in Hinged Racks

Perform pipetting accurately and consistently with these barrier pipette tips in hinged racks, which seal when exposed to potential contaminants. ART hinged racks are available in low retention, micropoint, extended length and other specialty styles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ModelFormatTip Style
2065E-05-HR100 μLART 100EHinged RackLow Retention
2139-HR10 μLART 10Hinged RackUniversal Fit
2139-05-HR10 μLART 10Hinged RackLow Retention
2149E-HR10 μLART 10 Ultra-microHinged RackUltra-micro
2149E-05-HR10 μLART 20E Ultra-microHinged RackLow Retention
2149P-HR20 μLART 20PHinged RackUniversal Fit
2149P-05-HR20 μLART 20PHinged RackLow Retention
2065-HR100 μLART 100Hinged RackUniversal Fit
2065E-HR100 μLART 100EHinged RackUniversal Fit
2069-HR200 μLART 200Hinged RackUniversal Fit
2069-05-HR200 μLART 200Hinged RackLow Retention
2070-HR300 μLART BarrierRackUniversal Fit
2070-05-HR300 μLART BarrierRackLow Retention
2179-HR1000 μLART BarrierHinged RackUniversal Fit
Catalog number 2065E-05-HR
Price (BRL)
-
Volume (Metric):
100 μL
Model:
ART 100E
Format:
Hinged Rack
Tip Style:
Low Retention
Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in low retention, micropoint, extended length, and other specialty styles.
Features:
  • Stackable hinged racks save space in your lab
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 100E, Filtered, Sterile, Hinged Rack
Length (English)2 in.
Length (Metric)5.07cm
ModelART 100E
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatHinged Rack
Product LineART
SterilitySterile
Tip StyleLow Retention
Volume (Metric)100 μL
Unit SizeCase of 4800