L(+)-Arginine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Arginine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP, EP and specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2066-0712 kg
2066-0950 kg
2066-061 kg
Catalog number 2066-07
Price (MXN)
-
Quantity:
12 kg
This product is GMP Manufactured and meets USP, JP, BP, EP and specifications.
This product is GMP Manufactured and meets USP, JP, BP, EP and specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point368°C (1013 hPa)
CAS74-79-3
Chemical Name or MaterialL(+)-Arginine
ColorColorless
Melting Point238°C
Molecular FormulaC6H14N4O2
PackagingPolyethylene pail, screw top
SynonymL-Arginine, H-L-Arg-OH, Arginine, (S)-(+)-Arginine, H-Arg-OH
GradeMulti-Compendial/USP
Molecular Weight (g/mol)174.2
Percent Purity98.5 to 101.5%
Quantity12 kg
Unit SizeEach