L(+)-Arginine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Arginine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP, EP and specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
2066-0950 kg
2066-0712 kg
2066-061 kg
Número de catálogo 2066-09
Precio (USD)
-
Cantidad:
50 kg
This product is GMP Manufactured and meets USP, JP, BP, EP and specifications.
This product is GMP Manufactured and meets USP, JP, BP, EP and specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición368°C (1013 hPa)
CAS74-79-3
Nombre del producto químico o materialL(+)-Arginine
ColorColorless
Punto de fusión238°C
Fórmula molecularC6H14N4O2
EnvasePolyethylene drum, open head
SinónimoL-Arginine, H-L-Arg-OH, Arginine, (S)-(+)-Arginine, H-Arg-OH
GradoMulticomponente/USP
Peso molecular (g/mol)174.2
Porcentaje de pureza98.5 to 101.5%
Cantidad50 kg
Unit SizeEach