Sodium carbonate, 99+%, for analysis, granular
Sodium carbonate, 99+%, for analysis, granular
Sodium carbonate, 99+%, for analysis, granular
Thermo Scientific Chemicals

Sodium carbonate, 99+%, for analysis, granular

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
5 kg
Packaging:
View all
Plastic Drum
Catalog number 206800050
Price (TWD)
-
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS497-19-8
IUPAC Namedisodium carbonate
Molecular FormulaCNa2O3
InChI KeyCDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (285°C)
Total nitrogen compounds=<0.002 % (as N)
Chloride (Cl)=<20 ppm
Arsenic (As)=<1 ppm
Phosphate (PO4)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only