Lithium borohydride, 95%
Lithium borohydride, 95%
Lithium borohydride, 95%
Thermo Scientific Chemicals

Lithium borohydride, 95%

CAS: 16949-15-8 | BH4Li | 21.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Catalog number 206810500
Price (USD)
542.65
Online Exclusive
603.00
Save 60.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
542.65
Online Exclusive
603.00
Save 60.35 (10%)
Each
Chemical Identifiers
CAS16949-15-8
IUPAC Namelithium(1+) boranuide
Molecular FormulaBH4Li
InChI KeyUUKMSDRCXNLYOO-UHFFFAOYSA-N
SMILES[Li+].[BH4-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=94 %
Appearance (Form)Powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only