ART™ Barrier Reload Insert Pipette Tips
ART™ Barrier Reload Insert Pipette Tips
ART™ Barrier Reload Insert Pipette Tips
ART™ Barrier Reload Insert Pipette Tips
ART™ Barrier Reload Insert Pipette Tips
Thermo Scientific™
ART™ Barrier Reload Insert Pipette Tips
Obtain accurate and consistent performance with these pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume (Metric)ModelTip StyleFit for Purpose
2069-05-RI200 μLART 200Low Retention, Micro PointSterile and PCR Ready
2065E-05-RI100 μLART 100ELow Retention, Micro PointSterile and PCR Ready
2065E-RI100 μLART 100EMicro PointSterile and PCR Ready
2065-RI100 μLART 100Micro PointSterile and PCR Ready
2069-RI200 μLART 200Micro PointSterile and PCR Ready
2139-05-RI10 μLART 10Low Retention, Micro PointSterile and PCR Ready
2139-RI10 μLART 10Micro PointSterile and PCR Ready
2149E-RI10 μLART 20EMicro PointSterile and PCR Ready
2149P-05-RI20 μLART 20PLow Retention, Micro PointSterile and PCR Ready
2149P-RI20 μLART 20PMicro PointSterile and PCR Ready
Número do catálogo 2069-05-RI
Preço (BRL)
-
Volume (Metric):
200 μL
Model:
ART 200
Tip Style:
Low Retention, Micro Point
Fit for Purpose:
Sterile and PCR Ready
Obtain accurate and consistent performance with Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips, which seal when exposed to potential contaminants. The ART inserts are environmentally friendly, providing individually packaged blister tubs that are made out of recycled plastic, designed to be used with the reusable ART hinged racks and creating the ART Reload System. The tips from the reload inserts clip securely into the hinged racks for efficient reloading.
Features
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in MicroPoint,extended length, low retention and SoftFit-L™ designed for use with Rainin™ LTS™ LiteTouch™ Pipettes
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Especificações
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Length (English)2.32 in.
Length (Metric)5.88cm
ModelART 200
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatRefill
Product LineART
SterilitySterile
Tip StyleLow Retention, Micro Point
Volume (Metric)200 μL
Unit SizeCase of 4800