Naringina, 97 %, Thermo Scientific Chemicals
Naringina, 97 %, Thermo Scientific Chemicals
Naringina, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Naringina, 97 %, Thermo Scientific Chemicals

CAS: 10236-47-2 | C27H32O14 | 580.54 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Envase:
Frasco de plástico
Número de catálogo 206910250
Precio (EUR)
67,70
Each
Cantidad:
25 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Precio (EUR)
67,70
Each
Identificadores químicos
CAS10236-47-2
IUPAC Name(2S)-7-{[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-2-yl]oxy}-5-hydroxy-2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Molecular FormulaC27H32O14
InChI KeyDFPMSGMNTNDNHN-ZPHOTFPESA-N
SMILESC[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Specific optical rotation-77° to -93° (20°C, 589 nm) (c=1, ethanol)
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only