ART™ Wide Bore Filtered Pipette Tips
Thermo Scientific™
ART™ Wide Bore Filtered Pipette Tips
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Fit for Purpose
2069G200 μLSterile and PCR Ready
2069GPK200 μLSterile and PCR Ready
2079G1000 μLSterile and PCR Ready
2079GPK1000 μLSterile and PCR Ready
Catalog number 2069G
Price (HKD)
-
Volume (Metric):
200 μL
Fit for Purpose:
Sterile and PCR Ready
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Wide Bore Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.
Features:
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
  • Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 200G, Filtered, Sterile, Lift-off Lid Rack
Length (English)1.96 in.
Length (Metric)4.97cm
ModelART 200G
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatLift-off Lid Rack
Product LineART
SterilitySterile
Tip StyleGenomic/Wide Orifice
Volume (Metric)200 μL
Unit SizeCase of 4800