4-Fluorocinnamic acid, 99%
4-Fluorocinnamic acid, 99%
4-Fluorocinnamic acid, 99%
Thermo Scientific Chemicals

4-Fluorocinnamic acid, 99%

CAS: 459-32-5 | C9H7FO2 | 166.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 207110250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS459-32-5
IUPAC Name(2E)-3-(4-fluorophenyl)prop-2-enoic acid
Molecular FormulaC9H7FO2
InChI KeyISMMYAZSUSYVQG-ZZXKWVIFSA-N
SMILESOC(=O)\C=C\C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point207°C to 210°C
Infrared spectrumConforms
Appearance (Form)Crystalline solid
HPLC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only