L(-)-Phenylephrine hydrochloride, 99%
L(-)-Phenylephrine hydrochloride, 99%
L(-)-Phenylephrine hydrochloride, 99%
Thermo Scientific Chemicals

L(-)-Phenylephrine hydrochloride, 99%

CAS: 61-76-7 | C9H14ClNO2 | 203.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 207241000
Price (USD)
456.65
Online exclusive
507.00
Save 50.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
456.65
Online exclusive
507.00
Save 50.35
Each
Chemical Identifiers
CAS61-76-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Specific optical rotation-43° to -47° (20°C, 589 nm) (c=2, H2O)
Infrared spectrumConforms
Appearance (Color)White to almost white
Melting point140°C to 145°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only