Spironolactone, 99%
Spironolactone, 99%
Spironolactone, 99%
Thermo Scientific Chemicals

Spironolactone, 99%

CAS: 52-01-7 | C24H32O4S | 416.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 207460010
Price (USD)
70.30
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
70.30
Each
Chemical Identifiers
CAS52-01-7
IUPAC Name(1R,3aS,3bR,4R,9aR,9bS,11aS)-4-(acetylsulfanyl)-9a,11a-dimethyl-2,3,3a,3b,4,5,7,8,9,9a,9b,10,11,11a-tetradecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione
Molecular FormulaC24H32O4S
InChI KeyLXMSZDCAJNLERA-ZHYRCANASA-N
SMILESCC(=O)S[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]2CC[C@@]3(C)[C@@H](CC[C@@]33CCC(=O)O3)[C@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (105°C, 2 h)
Specific optical rotation-41° to -45° (20°C, 589 nm) (c=1, EtOH)
HPLC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only