Squalene, 99+%, Thermo Scientific Chemicals
Squalene, 99+%, Thermo Scientific Chemicals
Squalene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Squalene, 99+%, Thermo Scientific Chemicals

Squalene, CAS # 111-02-4, is a biosynthetic precursor to all steroids and ergosterol and is used as an immunologic adjuvant in vaccines.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 207471000
Price (EUR)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-02-4
IUPAC Name(6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
Molecular FormulaC30H50
InChI KeyYYGNTYWPHWGJRM-AAJYLUCBSA-N
SMILESCC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.4945 to 1.4965 (20°C, 589 nm)
Density(20°C) 0.845 to 0.865 g/ml
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Squalene is a precursor to steroids and ergosterols
• It can be used in preparation of lipids, steroids, and fat-soluble vitamins

Applications
• Squalene is used as an immunologic adjuvant in vaccines
• It plays a vital role in the preparation of cholesterol, steroid hormones, and vitamin D
• It can also serve as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces

RUO – Research Use Only

General References:

  1. Sánchez-Fidalgo, S.; Villegas, I.; Rosillo, M. Á.; Aparicio-Soto, M.; de la Lastra, C. A. Dietary squalene supplementation improves DSS-induced acute colitis by downregulating p38 MAPK and NFkB signaling pathways. Mol. Nutr. Food Res. 2015, 59 (2), 284-292.
  2. Kedl, J. D.; Kedl, R. M. How squalene GLAdly helps generate antigen-specific T cells via antigen-carrying neutrophils and IL-18. Eur. J. Immunol. 2015, 45 (2), 376-379.