Tetramethylammonium hydroxide pentahydrate, 97%
Tetramethylammonium hydroxide pentahydrate, 97%
Tetramethylammonium hydroxide pentahydrate, 97%
Thermo Scientific Chemicals

Tetramethylammonium hydroxide pentahydrate, 97%

CAS: 10424-65-4 | C4H23NO6 | 181.23 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
產品號碼 207521000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS10424-65-4
IUPAC Nametetramethylazanium pentahydrate hydroxide
Molecular FormulaC4H23NO6
InChI KeyMYXKPFMQWULLOH-UHFFFAOYSA-M
SMILESO.O.O.O.O.[OH-].C[N+](C)(C)C
檢視更多
規格Specification Sheet規格表
Titration with HCl>=96.0 %
Appearance (Form)Hygroscopic powder or crystals
Melting point62°C to 73°C
Infrared spectrumConforms
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only