Nickel(II) chloride hexahydrate, 97%
Nickel(II) chloride hexahydrate, 97%
Nickel(II) chloride hexahydrate, 97%
Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 97%

CAS: 7791-20-0 | Cl2Ni·6H2O | 237.71 g/mol
数量:
500 g
2 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 207675000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7791-20-0
IUPAC Namenickel(2+) hexahydrate dichloride
Molecular FormulaCl2H12NiO6
InChI KeyLAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
さらに表示
pH4 to 7 (5% soln., 25°C)
Cobalt (Co)=<60 ppm
Iron (Fe)=<20 ppm
Zinc (Zn)=<10 ppm
Sodium (Na)=<50 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only