Copper(II) nitrate trihydrate, 99%, for analysis
Copper(II) nitrate trihydrate, 99%, for analysis
Copper(II) nitrate trihydrate, 99%, for analysis
Thermo Scientific Chemicals

Copper(II) nitrate trihydrate, 99%, for analysis

CAS: 10031-43-3 | CuN2O6·3H2O | 241.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 207680025
Price (USD)
369.65
Online exclusive
410.00
Save 40.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
369.65
Online exclusive
410.00
Save 40.35
Each
Chemical Identifiers
CAS10031-43-3
IUPAC Namecopper(2+) dinitrate
Molecular FormulaCuN2O6
InChI KeyXTVVROIMIGLXTD-UHFFFAOYSA-N
SMILES[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric98.5 to 104.0 %
Sulfate (SO4)=<50 ppm
Zinc (Zn)=<50 ppm
Appearance (Color)Blue
Appearance (Form)Powder or crystals and/or chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only