Copper, 99.9%, (trace metal basis), shot 1-10 mm
Copper, 99.9%, (trace metal basis), shot 1-10 mm
Copper, 99.9%, (trace metal basis), shot 1-10 mm
Thermo Scientific Chemicals

Copper, 99.9%, (trace metal basis), shot 1-10 mm

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
產品號碼 207691000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
檢視更多
規格Specification Sheet規格表
Total trace metal impurities=<0.1 %
Appearance (Form)Metallic shots
Particle size1 to 10 mm
Appearance (Color)Rust brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only