L(+)-Glutamic acid, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Glutamic acid, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, BP, and EP specifications.
Have Questions?
Catalog NumberQuantity
2077-061 kg
Catalog number 2077-06
Price (TWD)
-
Quantity:
1 kg
This product is GMP Manufactured and meets USP, BP, and EP specifications.
This product is GMP Manufactured and meets USP, BP, and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point325°C (1013 hPa)
CAS56-86-0
Chemical Name or MaterialL(+)-Glutamic acid
ColorWhite
Melting Point160°C
Molecular FormulaHO2CCH2CH2CH(NH2)CO2H
PackagingPoly bottle, tamper-evident cap
SynonymL-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid, H-Glu-OH, (S)-(+)-Glutamic acid
GradeMulti-Compendial/USP
Molecular Weight (g/mol)147.13
Percent Purity98.5 to 101.5%
Quantity1 kg
Unit SizeEach