Sodium peroxide, 96%
Sodium peroxide, 96%
Sodium peroxide, 96%
Thermo Scientific Chemicals

Sodium peroxide, 96%

CAS: 1313-60-6 | Na2O2 | 77.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 207701000
Price (USD)
127.65
Special offer
Online exclusive
Ends: 28-Dec-2026
150.00
Save 22.35
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
127.65
Special offer
Online exclusive
Ends: 28-Dec-2026
150.00
Save 22.35
Each
Chemical Identifiers
CAS1313-60-6
IUPAC Namedisodium dioxidanediide
Molecular FormulaNa2O2
InChI KeyPFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<50 ppm
Lead (Pb)=<50 ppm
Appearancematerial from the manufacturing process
Sulfate (SO4)=<50 ppm
Appearance (Color)Yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only