Oxalate de potassium monohydraté, 99+ %, pour analyse, Thermo Scientific Chemicals
Oxalate de potassium monohydraté, 99+ %, pour analyse, Thermo Scientific Chemicals
Oxalate de potassium monohydraté, 99+ %, pour analyse, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Oxalate de potassium monohydraté, 99+ %, pour analyse, Thermo Scientific Chemicals

CAS: 6487-48-5 | C2H2K2O5 | 184.23 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
500 g
2.5 kg
Conditionnement:
Flacon en plastique
Référence 207710025
Prix (EUR)
170,00
Each
Quantité:
2.5 kg
Conditionnement:
Flacon en plastique
Grand volume ou format personnalisé
Prix (EUR)
170,00
Each
Identifiants chimiques
CAS6487-48-5
IUPAC Namedipotassium hydrate oxalate
Molecular FormulaC2H2K2O5
InChI KeyQCPTVXCMROGZOL-UHFFFAOYSA-L
SMILESO.[K+].[K+].[O-]C(=O)C([O-])=O
Voir plus
Infrared spectrumConforms
Chloride (Cl)=<50 ppm
Iron (Fe)=<10 ppm
Appearance (Form)Crystals or flakes
Sulfate (SO4)=<100 ppm
Voir plus
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only