Sodium carbonate, 99.95%, extra pure, anhydrous
Sodium carbonate, 99.95%, extra pure, anhydrous
Sodium carbonate, 99.95%, extra pure, anhydrous
Thermo Scientific Chemicals

Sodium carbonate, 99.95%, extra pure, anhydrous

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 207765000
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
106.00
Save 15.35 (14%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
106.00
Save 15.35 (14%)
Each
Chemical Identifiers
CAS497-19-8
IUPAC Namedisodium carbonate
Molecular FormulaCNa2O3
InChI KeyCDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline or granular powder
Titration with HCl>=99.90 % (on dried substance)
Loss on drying=<1 % (285°C)
Heavy metals (as Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only