Potassium sulfate, 99+%, for analysis, anhydrous powder
Potassium sulfate, 99+%, for analysis, anhydrous powder
Potassium sulfate, 99+%, for analysis, anhydrous powder
Thermo Scientific Chemicals

Potassium sulfate, 99+%, for analysis, anhydrous powder

CAS: 7778-80-5 | K2O4S | 174.25 g/mol
Quantity:
500 g
5 kg
Packaging:
Plastic bottle
Catalog number 207770050
Price (JPY)
-
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7778-80-5
IUPAC Namedipotassium sulfate
Molecular FormulaK2O4S
InChI KeyOTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH5.5 to 8.0 (5 % at 20°C)
Nitrogen compounds (as N)=<0.0005 %
Lead (Pb)=<5 ppm
Chloride (Cl)=<10 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only