Specifications
Boiling Point355°C (1013 hPa)
CAS56-85-9
Chemical Name or MaterialL(+)-Glutamine
ColorWhite
Melting Point185°C to 186°C
Molecular FormulaC5H10N2O3
PackagingPoly bottle, tamper-evident cap
Synonym(S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamine, Levoglutamide, (S)-(+)-Glutamine, Gln
GradeMulti-Compendial
Molecular Weight (g/mol)146.15
Percent Purity98.5 to 101.5%
Quantity1 kg
pH4.5 to 6
Unit SizeEach