Sodium phosphate, monobasic monohydrate, 99+%, for analysis
Sodium phosphate, monobasic monohydrate, 99+%, for analysis
Sodium phosphate, monobasic monohydrate, 99+%, for analysis
Thermo Scientific Chemicals

Sodium phosphate, monobasic monohydrate, 99+%, for analysis

CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol
数量:
250 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 207802500
価格(JPY)
-
数量:
250 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10049-21-5
IUPAC Namesodium phosphoric acid hydrate
Molecular FormulaH5NaO5P
InChI KeyBBMHARZCALWXSL-UHFFFAOYSA-N
SMILESO.[Na+].OP(O)(O)=O
さらに表示
Insoluble matter=<0.01 % (Ca and NH4OH ppt)
Sulfate (SO4)=<30 ppm
Calcium (Ca)=<50 ppm
Appearance (Form)Crystalline powder or crystals
Heavy metals (as Pb)=<5 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only