Manganese(II) bromide, 99%, anhydrous
Manganese(II) bromide, 99%, anhydrous
Manganese(II) bromide, 99%, anhydrous
Thermo Scientific Chemicals

Manganese(II) bromide, 99%, anhydrous

CAS: 13446-03-2 | Br2Mn | 214.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 207860250
Price (USD)
112.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
112.00
Each
Chemical Identifiers
CAS13446-03-2
IUPAC Namemanganese(2+) dibromide
Molecular FormulaBr2Mn
InChI KeyRJYMRRJVDRJMJW-UHFFFAOYSA-L
SMILES[Mn++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric>=98.5 %
Appearance (Color)Pink
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only