Tris(triphenylphosphine)ruthenium(II) chloride, 98%
Tris(triphenylphosphine)ruthenium(II) chloride, 98%
Tris(triphenylphosphine)ruthenium(II) chloride, 98%
Thermo Scientific Chemicals

Tris(triphenylphosphine)ruthenium(II) chloride, 98%

CAS: 15529-49-4 | C54H45Cl2P3Ru | 958.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 207880250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS15529-49-4
IUPAC Nameλ¹-ruthenium(1+) tris(triphenylphosphane) dichloride
Molecular FormulaC54H45Cl2P3Ru
InChI KeyKSHWMFNJCNGTJO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ru+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Carbon (C)66.0 to 69.0 %
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Appearance (Color)Red-brown to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only