L(+)-Histidine, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Histidine, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
2080-061 kg
2080-05100 g
2080-0950 kg
2080-0712 kg
Número de catálogo 2080-06
Precio (MXN)
-
Cantidad:
1 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
CAS71-00-1
Nombre del producto químico o materialL(+)-Histidine
ColorWhite
Punto de fusión272°C to 273°C
Fórmula molecularC6H9N3O2
EnvasePoly bottle, tamper-evident cap
SinónimoL-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773, L-Histidine
GradoMulticomponente/USP
Peso molecular (g/mol)155.16
Porcentaje de pureza98.5 to 101.5%
Cantidad1 kg
pH7 to 8.5
Unit SizeEach